Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108791.1 | NP_001102261.1 | 4 | 1201 | ||
XM_006244673.2 | XP_006244735.1 | 3 | 887 | |||
XM_006244675.2 | XP_006244737.1 | 3 | 1054 | |||
XM_006244676.2 | XP_006244738.1 | 3 | 947 | |||
XM_006244677.2 | XP_006244739.1 | 4 | 871 | |||
XM_008766954.1 | XP_008765176.1 | 4 | 824 |