Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DDX3Y SIRNA Assays ›
Gene Symbol:
Ddx3y
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box helicase 3, Y-linked
Gene Aliases:
RGD1309586
Chromosome Location:
Chr.13: 105408179 - 105411348 on Build Rnor_6.0
UniGene ID:
Rn.218622
Species:
Rat
siRNA ID
RSS360937
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ddx3y
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001108858.1
NP_001102328.1
1
1220
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
chromosome segregation
metabolic process
extrinsic apoptotic signaling pathway via death domain receptors
response to virus
RNA secondary structure unwinding
positive regulation of gene expression
negative regulation of translation
positive regulation of cell growth
negative regulation of cell growth
negative regulation of protein complex assembly
DNA duplex unwinding
stress granule assembly
intracellular signal transduction
mature ribosome assembly
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of viral genome replication
innate immune response
positive regulation of translation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of translational initiation
cellular response to arsenic-containing substance
cellular response to osmotic stress
intrinsic apoptotic signaling pathway
positive regulation of G1/S transition of mitotic cell cycle
negative regulation of intrinsic apoptotic signaling pathway
nucleic acid binding
DNA binding
RNA binding
GTPase activity
ATP-dependent DNA helicase activity
ATP-dependent RNA helicase activity
helicase activity
ATP binding
transcription factor binding
poly(A) binding
eukaryotic initiation factor 4E binding
ATPase activity
nucleoside-triphosphatase activity
translation initiation factor binding
RNA stem-loop binding
ribosomal small subunit binding
CTPase activity
poly(A) RNA binding
mRNA 5'-UTR binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0