Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1564319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126290.1 | NP_001119762.1 | 1 | 122 | ||
GenBank mRNA | BC160905.1 | - | 1 | 122 | 7455398 |