Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765053.1 | XP_008763275.1 | 4 | 1172 | ||
XM_008765054.1 | XP_008763276.1 | 4 | 1167 | |||
XM_008765055.1 | XP_008763277.1 | 4 | 1156 | |||
XM_008765056.1 | XP_008763278.1 | 4 | 1172 | |||
GenBank mRNA | FQ218828.1 | - | 4 | 739 |