Search Thermo Fisher Scientific
Gene Symbol: Map4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024278.1 | NP_001019449.1 | 7 | 841 | ||
XM_006243813.2 | XP_006243875.1 | 7 | 865 | |||
XM_006243814.2 | XP_006243876.1 | 7 | 865 | |||
XM_006243815.2 | XP_006243877.1 | 7 | 865 | |||
XM_006243816.2 | XP_006243878.1 | 7 | 865 | |||
XM_006243818.2 | XP_006243880.1 | 7 | 866 | |||
XM_006243819.2 | XP_006243881.1 | 7 | 866 | |||
XM_006243820.2 | XP_006243882.1 | 7 | 867 | |||
XM_006243821.2 | XP_006243883.1 | 7 | 849 | |||
XM_006243822.2 | XP_006243884.1 | 7 | 849 | |||
XM_006243823.2 | XP_006243885.1 | 7 | 849 | |||
GenBank mRNA | BC088405.1 | - | 7 | 841 | 7110394 |