Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Praf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109013.1 | NP_001102483.1 | 2 | 517 | ||
GenBank mRNA | FQ212649.1 | - | 2 | 376 | ||
FQ213026.1 | - | 2 | 376 | |||
FQ213646.1 | - | 2 | 373 | |||
FQ219908.1 | - | 2 | 376 |