Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002288.1 | NP_001002288.1 | 1 | 132 | ||
XM_006230870.2 | XP_006230932.1 | 4 | 698 | |||
XM_006230872.2 | XP_006230934.1 | 2 | 496 | |||
XM_008760163.1 | XP_008758385.1 | 3 | 594 |
Set Membership: |
G-protein coupled receptor |