Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008899.1 | NP_001008899.1 | 2 | 710 | ||
XM_006242436.2 | XP_006242498.1 | 2 | 1214 |