Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008874.1 | NP_001008874.1 | 4 | 206 | ||
XM_003752044.1 | XP_003752092.1 | 4 | 206 |
Gene Symbol: LOC100910942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749193.1 | XP_003749241.1 | 4 | 206 |
Gene Symbol: LOC100911681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751772.2 | XP_003751820.1 | 4 | 206 |
Gene Symbol: LOC103690055 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772230.1 | XP_008770452.1 | 233 |