Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-CE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008833.1 | NP_001008833.1 | 3 | 485 | ||
XM_006255847.2 | XP_006255909.1 | 3 | 490 | |||
GenBank mRNA | BC083874.1 | - | 2 | 669 | ||
BC098952.1 | - | 3 | 488 | 7366301 |