Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jph4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003711.1 | NP_001003711.1 | 3 | 1242 | ||
XM_008770690.1 | XP_008768912.1 | 4 | 2040 | |||
GenBank mRNA | AY341260.1 | - | 3 | 1242 |