Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009539.1 | NP_001009539.1 | 4 | 745 | ||
XM_006250253.2 | XP_006250315.1 | 4 | 776 | |||
XM_006250254.2 | XP_006250316.1 | 4 | 896 | |||
GenBank mRNA | BC061812.1 | - | 4 | 745 | 5598658 |