Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Gsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009920.2 | NP_001009920.1 | 3 | 539 | ||
NM_001159739.2 | NP_001153211.1 | 4 | 509 | |||
NM_001287025.1 | NP_001273954.1 | 3 | 401 | |||
GenBank mRNA | BC100080.1 | - | 4 | 366 | 7324340 | |
CO566642.1 | - | 4 | 372 | |||
DY308874.1 | - | 4 | 433 | |||
DY573045.1 | - | 2 | 234 | |||
FQ210706.1 | - | 3 | 368 | |||
FQ218526.1 | - | 4 | 382 | |||
FQ218726.1 | - | 4 | 376 | |||
FQ219557.1 | - | 4 | 376 | |||
FQ219615.1 | - | 4 | 509 | |||
FQ219704.1 | - | 4 | 376 | |||
FQ231428.1 | - | 4 | 380 | |||
S72506.1 | - | 3 | 387 | |||
S82820.1 | - | 3 | 539 | |||
X78847.1 | - | 3 | 387 |