Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Apobec4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017492.1 | NP_001017492.1 | 2 | 747 | ||
XM_008769666.1 | XP_008767888.1 | 2 | 1554 | |||
XM_008769667.1 | XP_008767889.1 | 2 | 958 | |||
GenBank mRNA | BC079272.1 | - | 2 | 747 | 7134003 |