Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017494.1 | NP_001017494.1 | 3 | 221 | ||
GenBank mRNA | BC079114.1 | - | 3 | 221 | 7126053 |