Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PKD2 SIRNA Assays ›
Gene Symbol:
Pkd2
Gene Name:
polycystic kidney disease 2 (autosomal dominant)
Gene Aliases:
RGD1559992
Chromosome Location:
Chr.14: 6602004 - 6645257 on Build Rnor_6.0
UniGene ID:
Rn.198353
Species:
Rat
siRNA ID
RSS363543
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pkd2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001191934.1
NP_001178863.1
4, 5
1085
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
branching involved in ureteric bud morphogenesis
kidney development
liver development
embryonic placenta development
heart looping
detection of nodal flow
calcium ion transport
cellular calcium ion homeostasis
cell cycle arrest
positive regulation of cytosolic calcium ion concentration
JAK-STAT cascade
determination of left/right symmetry
heart development
positive regulation of gene expression
spinal cord development
neural tube development
regulation of cAMP metabolic process
positive regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
metanephric part of ureteric bud development
sodium ion transmembrane transport
aorta development
regulation of cell proliferation
cytoplasmic sequestering of transcription factor
positive regulation of nitric oxide biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
detection of mechanical stimulus
release of sequestered calcium ion into cytosol
transmembrane transport
negative regulation of ryanodine-sensitive calcium-release channel activity
placenta blood vessel development
renal tubule morphogenesis
renal artery morphogenesis
calcium ion transmembrane transport
positive regulation of cell cycle arrest
cellular response to hydrostatic pressure
cellular response to osmotic stress
cellular response to fluid shear stress
potassium ion transmembrane transport
determination of liver left/right asymmetry
renal system development
metanephric mesenchyme development
mesonephric tubule development
mesonephric duct development
metanephric smooth muscle tissue development
metanephric cortex development
metanephric ascending thin limb development
metanephric cortical collecting duct development
metanephric distal tubule development
metanephric S-shaped body morphogenesis
regulation of calcium ion import
negative regulation of G1/S transition of mitotic cell cycle
receptor binding
voltage-gated ion channel activity
voltage-gated calcium channel activity
voltage-gated sodium channel activity
calcium channel activity
potassium channel activity
calcium ion binding
cytoskeletal protein binding
channel activity
voltage-gated cation channel activity
identical protein binding
protein homodimerization activity
actinin binding
HLH domain binding
ion channel binding
calcium-induced calcium release activity
ATPase binding
phosphoprotein binding
muscle alpha-actinin binding
alpha-actinin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0