Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134609.2 | NP_001128081.2 | 5 | 694 | ||
XM_006228197.2 | XP_006228259.1 | 5 | 887 | |||
XM_006228199.2 | XP_006228261.1 | 5 | 884 |