Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100991.2 | NP_001094461.1 | 5 | 691 | ||
XM_006228733.2 | XP_006228795.1 | 2 | 3409 | |||
XM_006228735.2 | XP_006228797.1 | 4 | 4748 | |||
XM_006228737.2 | XP_006228799.1 | 4 | 3808 | |||
XM_006228738.2 | XP_006228800.1 | 5 | 3945 | |||
XM_006228740.2 | XP_006228802.1 | 4 | 1359 | |||
XM_006228743.2 | XP_006228805.1 | 4 | 456 | |||
XM_008759177.1 | XP_008757399.1 | 5 | 3107 | |||
GenBank mRNA | BC128772.1 | - | 5 | 716 | ||
BC166403.1 | - | 3 | 355 | 8363623 |
Gene Symbol: LOC102546354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255153.2 | XP_006255215.1 | 2 | 3413 | ||
XM_006255155.2 | XP_006255217.1 | 5 | 3110 | |||
XM_006255160.2 | XP_006255222.1 | 4 | 1362 | |||
XM_006255164.2 | XP_006255226.1 | 4 | 456 | |||
XM_008772333.1 | XP_008770555.1 | 4 | 4751 | |||
XM_008772334.1 | XP_008770556.1 | 5 | 4251 | |||
XM_008772335.1 | XP_008770557.1 | 5 | 3947 | |||
XM_008772336.1 | XP_008770558.1 | 4 | 3806 | |||
XM_008772337.1 | XP_008770559.1 | 4 | 4774 |