Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229814.2 | XP_006229876.1 | 18 | 2922 | ||
XM_006229817.2 | XP_006229879.1 | 17 | 2905 | |||
GenBank mRNA | BC168913.1 | - | 7 | 1254 |