Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrd1as1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166303.1 | NP_001159775.1 | 3 | 162 | ||
XM_006255877.2 | XP_006255939.1 | 3 | 1025 | |||
XM_008772699.1 | XP_008770921.1 | 3 | 243 | |||
GenBank mRNA | FQ213683.1 | - | 2 | 272 | ||
FQ214505.1 | - | 3 | 219 |