Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC499544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047944.1 | NP_001041409.1 | 6 | 591 | ||
GenBank mRNA | AY539905.1 | - | 6 | 591 |