Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109232.2 | NP_001102702.1 | 2 | 402 | ||
GenBank mRNA | BC168861.1 | - | 2 | 402 | 7129053 |