Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109246.1 | NP_001102716.1 | 3, 4 | 399 | ||
XM_008763075.1 | XP_008761297.1 | 3, 4 | 436 | |||
GenBank mRNA | BC168850.1 | - | 3, 4 | 366 | 8362904 |