Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC500300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024334.2 | NP_001019505.2 | 2 | 695 | ||
XM_006237196.2 | XP_006237258.1 | 2 | 2328 | |||
XM_006237197.2 | XP_006237259.1 | 2 | 2349 | |||
GenBank mRNA | AY931022.1 | - | 2 | 652 | ||
BC087112.1 | - | 2 | 695 | 7193534 |