Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109254.1 | NP_001102724.1 | 2 | 571 | ||
XM_006237536.2 | XP_006237598.1 | 2 | 610 |
Set Membership: |
![]() |