Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031663.1 | NP_001026833.1 | 9 | 846 | ||
XM_006237933.2 | XP_006237995.1 | 8 | 889 | |||
GenBank mRNA | BC079473.1 | - | 9 | 852 | ||
BC098713.1 | - | 9 | 846 | 7322407 |
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237930.2 | XP_006237992.1 | 25 | 9127 | ||
XM_006237931.2 | XP_006237993.1 | 24 | 9554 | |||
XM_008763590.1 | XP_008761812.1 | 26 | 9425 | |||
XM_008763591.1 | XP_008761813.1 | 18 | 8620 |