Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cactin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081447.1 | NP_001074916.1 | 9 | 1825 | ||
XM_008765124.1 | XP_008763346.1 | 9 | 2929 | |||
XM_008765125.1 | XP_008763347.1 | 9 | 2929 | |||
XM_008765126.1 | XP_008763348.1 | 9 | 2316 | |||
XM_008765127.1 | XP_008763349.1 | 9 | 2000 | |||
GenBank mRNA | AY934505.1 | - | 9 | 1825 | ||
BC166406.1 | - | 9 | 2455 | 9028620 |