Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109298.1 | NP_001102768.1 | 3 | 550 | ||
XM_008766647.1 | XP_008764869.1 | 3 | 550 | |||
XM_008766648.1 | XP_008764870.1 | 3 | 226 | |||
GenBank mRNA | BC168977.1 | - | 3 | 529 | 7453880 |