Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013135.2 | NP_001013153.2 | 5 | 562 | ||
XM_006233681.2 | XP_006233743.1 | 4 | 472 | |||
XM_006233682.2 | XP_006233744.1 | 4 | 391 | |||
XM_006233683.2 | XP_006233745.1 | 5 | 451 | |||
XM_006233684.2 | XP_006233746.1 | 3 | 304 | |||
XM_006233685.2 | XP_006233747.1 | 3 | 328 | |||
XM_006233686.2 | XP_006233748.1 | 5 | 541 | |||
GenBank mRNA | BC168170.1 | - | 3 | 334 | 9025284 |
Gene Symbol: LOC100360061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729626.3 | XP_002729672.1 | 378 |
Gene Symbol: LOC100363321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252792.1 | XP_006252854.1 | 1 | 327 |
Gene Symbol: LOC100909786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750168.3 | XP_003750216.1 | 1 | 378 |
Gene Symbol: RGD1562289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_576917.5 | XP_576917.1 | 1 | 396 |