Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025288.1 | NP_001020459.1 | 3, 4 | 417 | ||
GenBank mRNA | BC097356.1 | - | 3, 4 | 417 | 7442140 |
Gene Symbol: LOC100909986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752209.3 | XP_003752257.1 | 4, 5 | 662 | ||
XM_006257731.2 | XP_006257793.1 | 4, 5 | 662 |