Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd200r1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768755.1 | XP_008766977.1 | 7 | 1020 | ||
XM_008768756.1 | XP_008766978.1 | 7 | 976 | |||
XM_008768757.1 | XP_008766979.1 | 6 | 966 | |||
XM_008768758.1 | XP_008766980.1 | 6 | 974 |