Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135018.1 | NP_001128490.1 | 4, 5 | 479 | ||
XM_008765961.1 | XP_008764183.1 | 3, 4 | 1823 | |||
XM_008765962.1 | XP_008764184.1 | 3, 4 | 1831 | |||
XM_008765963.1 | XP_008764185.1 | 2, 3 | 1555 | |||
GenBank mRNA | BC079105.1 | - | 4, 5 | 479 |