Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APOB SIRNA Assays ›
Gene Symbol:
Apob
Gene Name:
apolipoprotein B
Gene Aliases:
Aa1064, Ac1-060, Apo B-100, ApoB-100, ApoB-48
Chromosome Location:
Chr.6: 33176826 - 33216381 on Build Rnor_6.0
UniGene ID:
Rn.33815
Species:
Rat
siRNA ID
RSS367405
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Apob
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019287.2
NP_062160.2
26
10428
Back To Top
More Information
Panther Classification:
Molecular Function -
apolipoprotein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
lipid metabolic process
triglyceride mobilization
lipid transport
spermatogenesis
nervous system development
cholesterol metabolic process
fertilization
response to virus
response to carbohydrate
post-embryonic development
response to organic substance
response to selenium ion
positive regulation of gene expression
positive regulation of macrophage derived foam cell differentiation
positive regulation of lipid storage
positive regulation of cholesterol storage
lipid catabolic process
triglyceride catabolic process
cholesterol transport
sperm motility
response to lipopolysaccharide
cholesterol efflux
low-density lipoprotein particle remodeling
low-density lipoprotein particle clearance
lipoprotein metabolic process
lipoprotein biosynthetic process
lipoprotein catabolic process
cholesterol homeostasis
lipoprotein transport
regulation of cholesterol biosynthetic process
artery morphogenesis
cellular response to tumor necrosis factor
cellular response to prostaglandin stimulus
lipid transporter activity
protein binding
phospholipid binding
heparin binding
lipid binding
cholesterol transporter activity
lipase binding
low-density lipoprotein particle receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline