Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019304.1 | NP_062177.1 | 24 | 2451 | ||
GenBank mRNA | D16100.1 | - | 24 | 2451 |