Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019370.2 | NP_062243.2 | 18 | 1673 | ||
GenBank mRNA | BC097326.1 | - | 18 | 1673 | 7458431 | |
D30649.1 | - | 18 | 1700 | |||
FQ218696.1 | - | 1 | 58 | |||
U78787.1 | - | 18 | 1670 | |||
U78788.1 | - | 18 | 1676 | |||
Z47987.1 | - | 18 | 1657 |