Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYL1 SIRNA Assays ›
Gene Symbol:
Myl1
Gene Name:
myosin, light chain 1
Gene Aliases:
MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
Chromosome Location:
Chr.9: 73937820 - 73958480 on Build Rnor_6.0
UniGene ID:
Rn.84920
Species:
Rat
siRNA ID
RSS367544
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Myl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077656.1
NP_001071124.1
3
303
NM_020104.2
NP_064489.1
3
121
XM_008767294.1
XP_008765516.1
2
251
XM_008767295.1
XP_008765517.1
2
372
GenBank mRNA
CR755322.1
-
3
126
CR756356.1
-
3
121
FQ214580.1
-
3
430
FQ214604.1
-
3
348
FQ214608.1
-
3
308
FQ214631.1
-
3
290
FQ214638.1
-
3
397
FQ214656.1
-
3
139
FQ214675.1
-
3
397
FQ214712.1
-
3
311
FQ214723.1
-
3
311
FQ214727.1
-
3
311
FQ214742.1
-
3
311
FQ214939.1
-
3
308
FQ215002.1
-
3
397
FQ215050.1
-
3
311
FQ215083.1
-
3
308
FQ215086.1
-
3
141
FQ215201.1
-
3
308
FQ215229.1
-
3
402
FQ215325.1
-
3
397
FQ215340.1
-
3
139
FQ215418.1
-
3
397
FQ215435.1
-
3
308
FQ215492.1
-
3
290
FQ215500.1
-
2
887
FQ215535.1
-
2
372
FQ215557.1
-
3
397
FQ215563.1
-
3
308
FQ215578.1
-
3
397
FQ215610.1
-
3
311
FQ215748.1
-
2
370
FQ215766.1
-
3
397
FQ215774.1
-
3
308
FQ215848.1
-
3
308
FQ215875.1
-
3
308
FQ215911.1
-
3
402
FQ215933.1
-
3
311
FQ216034.1
-
3
311
FQ216060.1
-
3
397
FQ216109.1
-
3
308
FQ216124.1
-
3
402
FQ216181.1
-
3
290
FQ216213.1
-
4
305
FQ216218.1
-
3
308
FQ216229.1
-
3
578
FQ216238.1
-
3
397
FQ216308.1
-
3
139
FQ216407.1
-
3
479
FQ216452.1
-
3
308
FQ216455.1
-
3
312
FQ216484.1
-
3
290
FQ216496.1
-
3
311
FQ216525.1
-
3
311
FQ216543.1
-
3
308
FQ216556.1
-
3
397
FQ216567.1
-
3
308
FQ216631.1
-
3
311
FQ216654.1
-
3
395
FQ216704.1
-
3
311
FQ216737.1
-
3
394
FQ216738.1
-
3
139
FQ217074.1
-
3
138
FQ217103.1
-
3
139
FQ217157.1
-
2
114
FQ217192.1
-
3
139
FQ217218.1
-
3
139
FQ217243.1
-
3
138
FQ217303.1
-
3
290
FQ217323.1
-
3
139
FQ217387.1
-
3
139
FQ217394.1
-
3
241
FQ217427.1
-
3
144
FQ217481.1
-
3
139
FQ217524.1
-
3
139
FQ217753.1
-
3
144
FQ217756.1
-
3
139
FQ217889.1
-
3
139
FQ217973.1
-
3
144
FQ217979.1
-
3
139
FQ218029.1
-
3
139
FQ218049.1
-
3
139
FQ223584.1
-
3
397
FQ223632.1
-
3
311
FQ223641.1
-
3
397
FQ223687.1
-
3
139
FQ223779.1
-
3
139
FQ223827.1
-
3
311
FQ223880.1
-
3
209
FQ223984.1
-
3
139
FQ224047.1
-
3
139
FQ224050.1
-
3
139
FQ224087.1
-
3
139
FQ224112.1
-
2
114
FQ224155.1
-
3
144
FQ224171.1
-
3
308
FQ224172.1
-
3
139
FQ224212.1
-
3
291
FQ224277.1
-
3
311
FQ224321.1
-
3
143
FQ224386.1
-
3
144
FQ224411.1
-
3
139
FQ224420.1
-
3
139
FQ224452.1
-
3
214
FQ224519.1
-
3
308
FQ224609.1
-
4
167
FQ224663.1
-
3
139
FQ224726.1
-
3
241
FQ224796.1
-
3
139
FQ224799.1
-
3
144
FQ224927.1
-
3
397
FQ224931.1
-
3
308
FQ224934.1
-
3
306
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cardiac muscle contraction
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qvdz4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0