Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYL1 SIRNA Assays ›
Gene Symbol:
Myl1
Gene Name:
myosin, light chain 1
Gene Aliases:
MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
Chromosome Location:
Chr.9: 73937820 - 73958480 on Build Rnor_6.0
UniGene ID:
Rn.84920
Species:
Rat
siRNA ID
RSS367545
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Myl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077656.1
NP_001071124.1
3
355
NM_020104.2
NP_064489.1
3
173
XM_008767294.1
XP_008765516.1
2
303
XM_008767295.1
XP_008765517.1
2
424
GenBank mRNA
BI277595.1
-
5
632
CR755322.1
-
3
178
CR756356.1
-
3
173
FQ214580.1
-
3
482
FQ214604.1
-
3
400
FQ214608.1
-
3
360
FQ214631.1
-
3
342
FQ214638.1
-
3
449
FQ214656.1
-
3
191
FQ214675.1
-
3
449
FQ214712.1
-
3
363
FQ214723.1
-
3
363
FQ214727.1
-
3
363
FQ214742.1
-
3
363
FQ214939.1
-
3
360
FQ215002.1
-
3
449
FQ215050.1
-
3
363
FQ215086.1
-
3
193
FQ215201.1
-
3
360
FQ215229.1
-
3
454
FQ215325.1
-
3
449
FQ215340.1
-
3
191
FQ215418.1
-
3
449
FQ215435.1
-
3
360
FQ215492.1
-
3
342
FQ215500.1
-
2
939
FQ215535.1
-
2
424
FQ215557.1
-
3
449
FQ215563.1
-
3
360
FQ215578.1
-
3
449
FQ215610.1
-
3
363
FQ215748.1
-
2
422
FQ215766.1
-
3
449
FQ215774.1
-
3
360
FQ215820.1
-
3
418
FQ215848.1
-
3
360
FQ215875.1
-
3
360
FQ215911.1
-
3
454
FQ215933.1
-
3
363
FQ216034.1
-
3
363
FQ216060.1
-
3
449
FQ216109.1
-
3
360
FQ216124.1
-
3
454
FQ216181.1
-
3
342
FQ216192.1
-
3
449
FQ216213.1
-
4
357
FQ216218.1
-
3
360
FQ216229.1
-
3
630
FQ216238.1
-
3
449
FQ216308.1
-
3
191
FQ216407.1
-
3
531
FQ216452.1
-
3
360
FQ216455.1
-
3
364
FQ216484.1
-
3
342
FQ216496.1
-
3
363
FQ216525.1
-
3
363
FQ216543.1
-
3
360
FQ216556.1
-
3
449
FQ216567.1
-
3
360
FQ216631.1
-
3
363
FQ216654.1
-
3
447
FQ216704.1
-
3
363
FQ216737.1
-
3
446
FQ216738.1
-
3
191
FQ217074.1
-
3
190
FQ217103.1
-
3
191
FQ217157.1
-
2
166
FQ217192.1
-
3
191
FQ217218.1
-
3
191
FQ217243.1
-
3
190
FQ217303.1
-
3
342
FQ217323.1
-
3
191
FQ217387.1
-
3
191
FQ217394.1
-
3
293
FQ217427.1
-
3
196
FQ217481.1
-
3
191
FQ217524.1
-
3
191
FQ217753.1
-
3
196
FQ217756.1
-
3
191
FQ217889.1
-
3
191
FQ217973.1
-
3
196
FQ217979.1
-
3
191
FQ218029.1
-
3
191
FQ218049.1
-
3
191
FQ223584.1
-
3
449
FQ223632.1
-
3
363
FQ223687.1
-
3
191
FQ223755.1
-
3
359
FQ223779.1
-
3
191
FQ223827.1
-
3
363
FQ223880.1
-
3
261
FQ223949.1
-
3
189
FQ223984.1
-
3
191
FQ224047.1
-
3
191
FQ224050.1
-
3
191
FQ224087.1
-
3
191
FQ224112.1
-
2
166
FQ224155.1
-
3
196
FQ224171.1
-
3
360
FQ224172.1
-
3
191
FQ224212.1
-
3
343
FQ224254.1
-
3
191
FQ224277.1
-
3
363
FQ224321.1
-
3
195
FQ224386.1
-
3
196
FQ224411.1
-
3
191
FQ224420.1
-
3
191
FQ224452.1
-
3
266
FQ224519.1
-
3
360
FQ224609.1
-
4
219
FQ224663.1
-
3
191
FQ224726.1
-
3
293
FQ224796.1
-
3
191
FQ224799.1
-
3
196
FQ224867.1
-
3
449
FQ224927.1
-
3
449
FQ224931.1
-
3
360
FQ224934.1
-
3
358
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cardiac muscle contraction
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f4tdd:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0