Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077656.1 | NP_001071124.1 | 3, 4 | 413 | ||
NM_020104.2 | NP_064489.1 | 3, 4 | 231 | |||
XM_008767294.1 | XP_008765516.1 | 2, 3 | 361 | |||
XM_008767295.1 | XP_008765517.1 | 2, 3 | 482 | |||
GenBank mRNA | BI277595.1 | - | 4, 5 | 575 | ||
CR755322.1 | - | 3, 4 | 236 | |||
CR756356.1 | - | 3, 4 | 231 | |||
FQ214580.1 | - | 3, 4 | 540 | |||
FQ214608.1 | - | 3, 4 | 418 | |||
FQ214631.1 | - | 3, 4 | 400 | |||
FQ214638.1 | - | 3, 4 | 507 | |||
FQ214656.1 | - | 3, 4 | 249 | |||
FQ214675.1 | - | 3, 4 | 507 | |||
FQ214712.1 | - | 3, 4 | 421 | |||
FQ214723.1 | - | 3, 4 | 421 | |||
FQ214727.1 | - | 3, 4 | 421 | |||
FQ214742.1 | - | 3, 4 | 421 | |||
FQ214939.1 | - | 3, 4 | 418 | |||
FQ215002.1 | - | 3, 4 | 507 | |||
FQ215050.1 | - | 3, 4 | 421 | |||
FQ215083.1 | - | 3, 4 | 418 | |||
FQ215086.1 | - | 3, 4 | 251 | |||
FQ215201.1 | - | 3, 4 | 418 | |||
FQ215229.1 | - | 3, 4 | 512 | |||
FQ215325.1 | - | 3, 4 | 507 | |||
FQ215340.1 | - | 3, 4 | 249 | |||
FQ215418.1 | - | 3, 4 | 507 | |||
FQ215435.1 | - | 3, 4 | 418 | |||
FQ215492.1 | - | 3, 4 | 400 | |||
FQ215557.1 | - | 3, 4 | 507 | |||
FQ215563.1 | - | 3, 4 | 418 | |||
FQ215578.1 | - | 3, 4 | 507 | |||
FQ215610.1 | - | 3, 4 | 421 | |||
FQ215748.1 | - | 2, 3 | 480 | |||
FQ215766.1 | - | 3, 4 | 507 | |||
FQ215774.1 | - | 3, 4 | 418 | |||
FQ215820.1 | - | 3, 4 | 476 | |||
FQ215848.1 | - | 3, 4 | 418 | |||
FQ215875.1 | - | 3, 4 | 418 | |||
FQ215911.1 | - | 3, 4 | 512 | |||
FQ215933.1 | - | 3, 4 | 421 | |||
FQ216034.1 | - | 3, 4 | 421 | |||
FQ216060.1 | - | 3, 4 | 507 | |||
FQ216109.1 | - | 3, 4 | 418 | |||
FQ216124.1 | - | 3, 4 | 512 | |||
FQ216181.1 | - | 3, 4 | 400 | |||
FQ216192.1 | - | 3, 4 | 507 | |||
FQ216213.1 | - | 4, 5 | 415 | |||
FQ216218.1 | - | 3, 4 | 418 | |||
FQ216229.1 | - | 3, 4 | 688 | |||
FQ216238.1 | - | 3, 4 | 507 | |||
FQ216308.1 | - | 3, 4 | 249 | |||
FQ216407.1 | - | 3, 4 | 589 | |||
FQ216452.1 | - | 3, 4 | 418 | |||
FQ216455.1 | - | 3, 4 | 422 | |||
FQ216484.1 | - | 3, 4 | 400 | |||
FQ216496.1 | - | 3, 4 | 421 | |||
FQ216525.1 | - | 3, 4 | 421 | |||
FQ216543.1 | - | 3, 4 | 418 | |||
FQ216556.1 | - | 3, 4 | 507 | |||
FQ216567.1 | - | 3, 4 | 418 | |||
FQ216631.1 | - | 3, 4 | 421 | |||
FQ216704.1 | - | 3, 4 | 421 | |||
FQ216737.1 | - | 3, 4 | 504 | |||
FQ216738.1 | - | 3, 4 | 249 | |||
FQ217074.1 | - | 3, 4 | 248 | |||
FQ217103.1 | - | 3, 4 | 249 | |||
FQ217157.1 | - | 2, 3 | 224 | |||
FQ217192.1 | - | 3, 4 | 249 | |||
FQ217218.1 | - | 3, 4 | 249 | |||
FQ217243.1 | - | 3, 4 | 248 | |||
FQ217303.1 | - | 3, 4 | 400 | |||
FQ217323.1 | - | 3, 4 | 249 | |||
FQ217387.1 | - | 3, 4 | 249 | |||
FQ217394.1 | - | 3, 4 | 351 | |||
FQ217427.1 | - | 3, 4 | 254 | |||
FQ217481.1 | - | 3, 4 | 249 | |||
FQ217524.1 | - | 3, 4 | 249 | |||
FQ217753.1 | - | 3, 4 | 254 | |||
FQ217756.1 | - | 3, 4 | 249 | |||
FQ217889.1 | - | 3, 4 | 249 | |||
FQ217973.1 | - | 3, 4 | 254 | |||
FQ217979.1 | - | 3, 4 | 249 | |||
FQ218029.1 | - | 3, 4 | 249 | |||
FQ218049.1 | - | 3, 4 | 249 | |||
FQ223584.1 | - | 3, 4 | 507 | |||
FQ223632.1 | - | 3, 4 | 421 | |||
FQ223641.1 | - | 3, 4 | 507 | |||
FQ223687.1 | - | 3, 4 | 249 | |||
FQ223755.1 | - | 3, 4 | 417 | |||
FQ223779.1 | - | 3, 4 | 249 | |||
FQ223827.1 | - | 3, 4 | 421 | |||
FQ223880.1 | - | 3, 4 | 319 | |||
FQ223949.1 | - | 3, 4 | 247 | |||
FQ223984.1 | - | 3, 4 | 249 | |||
FQ224047.1 | - | 3, 4 | 249 | |||
FQ224050.1 | - | 3, 4 | 249 | |||
FQ224087.1 | - | 3, 4 | 249 | |||
FQ224112.1 | - | 2, 3 | 224 | |||
FQ224155.1 | - | 3, 4 | 254 | |||
FQ224171.1 | - | 3, 4 | 418 | |||
FQ224172.1 | - | 3, 4 | 249 | |||
FQ224254.1 | - | 3, 4 | 249 | |||
FQ224277.1 | - | 3, 4 | 421 | |||
FQ224321.1 | - | 3, 4 | 253 | |||
FQ224386.1 | - | 3, 4 | 254 | |||
FQ224411.1 | - | 3, 4 | 249 | |||
FQ224420.1 | - | 3, 4 | 249 | |||
FQ224452.1 | - | 3, 4 | 324 | |||
FQ224519.1 | - | 3, 4 | 418 | |||
FQ224609.1 | - | 4, 5 | 277 | |||
FQ224663.1 | - | 3, 4 | 249 | |||
FQ224726.1 | - | 3, 4 | 351 | |||
FQ224796.1 | - | 3, 4 | 249 | |||
FQ224799.1 | - | 3, 4 | 254 | |||
FQ224867.1 | - | 3, 4 | 507 | |||
FQ224927.1 | - | 3, 4 | 507 | |||
FQ224931.1 | - | 3, 4 | 418 | |||
FQ224934.1 | - | 3, 4 | 416 |