Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cacna1i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020084.3 | NP_064469.3 | 11, 12 | 2212 | ||
GenBank mRNA | AF086827.2 | - | 2477 | |||
AF290214.4 | - | 11, 12 | 2212 |