Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270586.1 | NP_001257515.1 | 9, 10 | 1234 | ||
NM_001270587.1 | NP_001257516.1 | 8, 9 | 1135 | |||
NM_031650.3 | NP_113838.1 | 9, 10 | 1234 | |||
GenBank mRNA | AF147740.1 | - | 8, 9 | 1120 | ||
AF208545.2 | - | 9, 10 | 1234 | |||
AF217450.2 | - | 8, 9 | 1135 | |||
AJ271682.1 | - | 9, 10 | 1235 |