Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbil5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021596.2 | NP_067607.1 | 2 | 336 | ||
GenBank mRNA | AF128531.1 | - | 2 | 338 |