Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021669.2 | NP_067701.1 | 3 | 274 | ||
GenBank mRNA | AB029433.1 | - | 3 | 275 | ||
AB035699.1 | - | 3 | 268 | |||
BF543543.1 | - | 3 | 270 |