Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kcnk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022293.1 | NP_071629.1 | 2 | 770 | ||
XM_006240447.2 | XP_006240509.1 | 3 | 3568 | |||
GenBank mRNA | AF287301.1 | - | 2 | 770 |