Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022400.1 | NP_071795.1 | 3 | 174 | ||
XM_006229149.1 | XP_006229211.1 | 3 | 351 | |||
XM_008759413.1 | XP_008757635.1 | 3 | 916 | |||
GenBank mRNA | BC061790.1 | - | 3 | 170 | 5623666 | |
U68417.1 | - | 3 | 174 |