Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022536.2 | NP_071981.1 | 1, 2 | 136 | ||
GenBank mRNA | AF071225.1 | - | 1, 2 | 136 | ||
BC061971.1 | - | 1, 2 | 165 | 5623342 | ||
FQ211358.1 | - | 1, 2 | 140 | |||
FQ212031.1 | - | 1, 2 | 165 | |||
FQ219792.1 | - | 1, 2 | 174 | |||
FQ219926.1 | - | 1, 2 | 186 | |||
FQ221020.1 | - | 1, 2 | 189 | |||
FQ221248.1 | - | 1, 2 | 184 | |||
FQ221362.1 | - | 1, 2 | 163 | |||
FQ222696.1 | - | 1, 2 | 298 | |||
FQ228087.1 | - | 1, 2 | 179 | |||
FQ229174.1 | - | 1, 2 | 186 | |||
FQ229214.1 | - | 1, 2 | 180 | |||
FQ230004.1 | - | 190 | ||||
FQ231656.1 | - | 1, 2 | 179 | |||
FQ232077.1 | - | 1, 2 | 175 | |||
FQ232920.1 | - | 1, 2 | 189 |