Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022615.1 | NP_072137.1 | 3, 4 | 166 | ||
GenBank mRNA | AF140782.1 | - | 3, 4 | 166 |
Gene Symbol: LOC499179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759598.1 | XP_008757820.1 | 3 | 304 |