Gene Symbol: Map1lc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022867.2 | NP_074058.2 | 3 | 261 | ||
GenBank mRNA | AY206669.1 | - | 3 | 223 | ||
AY310156.1 | - | 789 | ||||
AY392036.1 | - | 3 | 261 | |||
BC058144.1 | - | 3 | 219 | 6889616 | ||
BC083556.1 | - | 3 | 241 | 7189855 | ||
FQ221245.1 | - | 3 | 272 | |||
FQ228912.1 | - | 3 | 272 | |||
U05784.1 | - | 3 | 211 |