Search Thermo Fisher Scientific
Gene Symbol: Cldn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031701.2 | NP_113889.1 | 1 | 772 | ||
GenBank mRNA | BC082073.1 | - | 1 | 770 | 7133096 | |
FQ213419.1 | - | 1 | 786 |