Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031722.2 | NP_113910.1 | 4 | 577 | ||
XM_006249327.2 | XP_006249389.1 | 5 | 647 | |||
GenBank mRNA | BC062036.1 | - | 4 | 577 | 5623841 | |
X92097.1 | - | 4 | 548 |