Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044276.1 | NP_001037741.1 | 1 | 489 | ||
XM_008761775.1 | XP_008759997.1 | 2 | 2038 | |||
GenBank mRNA | BC091400.1 | - | 1 | 489 | 7323940 |